Ethyl-3-bromoimidazo[1,2-a]pyridine-2-carboxylate 97%

Warning

Synonyms: Ethyl 3-bromoimidazo[1,2-a]pyridine-2-carboxylate

A143905-5G A143905-1G
103834-382EA 72.69 USD
103834-382 103803-344
Ethyl-3-bromoimidazo[1,2-a]pyridine-2-carboxylate 97%
Ethyl 3-bromoimidazo[1,2-a]pyridine-2-carboxylate

Caution: Inert atmosphere, 2…8 °C.

Formula: C₁₀H₉BrN₂O₂
MW: 269.1000 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD05864806
CAS Number: 143982-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR