Ethyl-2-amino-4-methylthiophene-3-carboxylate 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Ethyl 2-amino-4-methylthiophene-3-carboxylate
A195399-1G
A195399-5G
103816-750EA
7
USD
103816-750
103847-364
Ethyl-2-amino-4-methylthiophene-3-carboxylate 98%
Ethyl 2-amino-4-methylthiophene-3-carboxylate
Caution: Keep in a dark place, sealed in dry, room temperature
Formula:
C₈H₁₁NO₂S MW: 185.25 g/mol Boiling Pt: 279.1 °C (760 mm Hg) Melting Pt: 72 °C Flash Pt: 122.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051669 CAS Number: 43088-42-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...