Diphenylphosphine oxide 97%

Supplier: Ambeed

Warning

A406801-5G
103838-182EA 8 USD
103838-182
Diphenylphosphine oxide 97%
Diphenylphosphine oxide

Caution: Inert atmosphere, room temperature

Formula: C₁₂H₁₁OP
Boiling Pt: 329.2±25.0 °C (760 mm Hg)
Melting Pt: 50…57 °C
Flash Pt: 152.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00002079
CAS Number: 4559-70-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR