Denatonium benzoate 99%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-Benzyl-2-((2,6-dimethylphenyl)amino)-N,N-diethyl-2-oxoethanaminium benzoate
A181673-10G
A181673-5G
103846-014EA
12.69
USD
103846-014
76690-590
Denatonium benzoate 99%
Denatonium benzoate
Caution: for Research Use Only
Formula:
C₂₈H₃₄N₂O₃ MW: 446.59 g/mol Melting Pt: 164…168 °C Storage Temperature: Ambient |
MDL Number:
MFCD00031578 CAS Number: 3734-33-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...