Chloro(diphenyl)methane 95%

Supplier: Ambeed
Danger

Synonyms: Benzhydryl chloride

A674425-5G A674425-5G A674425-100G
76844-714EA 6.18 USD
76844-714 76697-158 103840-516
Chloro(diphenyl)methane 95%
Chloro(diphenyl)methane

Caution: for Research Use Only

Formula: (C₆H₅)₂CHCl
MW: 202.68 g/mol
Boiling Pt: 315.5 °C (760 mm Hg)
Melting Pt: 15…17 °C
Flash Pt: 112 °C
Storage Temperature: Ambient
MDL Number: MFCD00000855
CAS Number: 90-99-3
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR