Isoquinoline-8-carbaldehyde 97%
Supplier: Ambeed
|
Warning
|
A218034-1G
A218034-1G
A218034-5G
77088-070EA
73.13
USD
77088-070
103818-682
77112-660
Isoquinoline-8-carbaldehyde 97%
Isoquinoline-8-carbaldehyde
Caution: for Research Use Only
Formula:
C₁₀H₇NO MW: 157.1687 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD06227440 CAS Number: 787615-01-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...