8-Bromoquinoline 98%

Supplier: Ambeed

Warning

A297506-1G A297506-5G
103835-280EA 12 USD
103835-280 103814-138
8-Bromoquinoline 98%
8-Bromoquinoline

Caution: Inert atmosphere, 2…8 °C.

Formula: C₉H₆BrN
MW: 208.06 g/mol
Boiling Pt: 303 °C (760 mm Hg)
Melting Pt: 58…59 °C
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00191859
CAS Number: 16567-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR