3-Amino-7-bromoisoquinoline 96%

Supplier: Ambeed

Warning

Synonyms: 7-Bromoisoquinolin-3-amine

A283286-5G A283286-1G
103813-606EA 389 USD
103813-606 103853-558
3-Amino-7-bromoisoquinoline 96%
3-Amino-7-bromoisoquinoline

Caution: Keep in a dark place, inert atmosphere, 2…8 °C

Formula: C₉H₇BrN₂
MW: 223.0693 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11040207
CAS Number: 1192815-02-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR