6-Nitrophthalide 98%

Supplier: Ambeed
Warning

Synonyms: 6-Nitroisobenzofuran-1(3H)-one

A377148-25G A377148-5G A377148-5G A377148-1G A377148-10G A377148-1G
103837-630EA 21.92 USD
103837-630 76840-936 77306-892 77306-894 77240-326 103826-602
6-Nitrophthalide 98%
6-Nitrophthalide

Caution: for Research Use Only

Formula: C₈H₅NO₄
MW: 179.13 g/mol
Boiling Pt: 413.4 °C (760 mm Hg)
Melting Pt: 140…145 °C
Flash Pt: 229.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00033529
CAS Number: 610-93-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR