2-Chloro-5-formylpyridine ≥95%

Supplier: Ambeed

Warning

Synonyms: 6-Chloronicotinaldehyde6-Chloropyridine-3-carboxaldehyde2-Chloropyridine-5-carboxaldehyde

A120702-5G A120702-1G
103830-798EA 14 USD
103830-798 103799-984
2-Chloro-5-formylpyridine ≥95%
2-Chloro-5-formylpyridine

Caution: Keep in a dark place, sealed in dry, room temperature

Formula: C₆H₄ClNO
MW: 141.56 g/mol
Boiling Pt: 249.3 °C (760 mm Hg)
Melting Pt: 77…81 °C
Flash Pt: 104.6 °C
Storage Temperature: Ambient
MDL Number: MFCD03095223
CAS Number: 23100-12-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR