6-Bromoquinoxaline 98%

Supplier: Ambeed

Warning

A451473-5G A451473-1G
103827-792EA 9 USD
103827-792 103838-862
6-Bromoquinoxaline 98%
6-Bromoquinoxaline

Caution: Sealed in dry, at room temperature.

Formula: C₈H₅BrN₂
MW: 209.0427 g/mol
Boiling Pt: 300.2 °C (760 mm Hg)
Melting Pt: 54 °C
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD00837757
CAS Number: 50998-17-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR