6-Bromoimidazo[1,2-a]pyridine 97%

Supplier: Ambeed
Warning

A149665-5G A149665-500G A149665-25G A149665-1G A149665-5G A149665-10G A149665-1G A149665-100G
77317-694EA 13 USD
77317-694 103842-192 76812-148 76812-146 77299-746 77299-748 76812-150 103804-066
6-Bromoimidazo[1,2-a]pyridine 97%
6-Bromoimidazo[1,2-a]pyridine

Caution: for Research Use Only

Formula: C₇H₅BrN₂
MW: 197.03 g/mol
Melting Pt: 76…78 °C
Storage Temperature: Ambient
MDL Number: MFCD06496200
CAS Number: 6188-23-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR