4,5-Dimethoxyisatoic anhydride 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
6,7-Dimethoxy-1H-benzo[d][1,3]oxazine-2,4-dione
A114213-1G
A114213-5G
A114213-250MG
A114213-1G
76766-048EA
14.42
USD
76766-048
77311-030
103798-372
103824-212
4,5-Dimethoxyisatoic anhydride 97%
4,5-Dimethoxyisatoic anhydride
Caution: for Research Use Only
Formula:
C₁₀H₉NO₅ MW: 223.1800 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03426390 CAS Number: 20197-92-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...