5-Fluoro-8-quinolinol 95%

Supplier: Ambeed

Warning

Synonyms: 5-Fluoro-8-hydroxyquinoline

A542959-1G
103828-646EA 294 USD
103828-646
5-Fluoro-8-quinolinol 95%
5-Fluoro-8-quinolinol

Caution: Inert atmosphere, room temperature

Formula: C₉H₆FNO
Boiling Pt: 222.4 °C (760 mm Hg)
Flash Pt: 73.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00059754
CAS Number: 387-97-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR