5-Fluoro-3-iodo-1H-indazole 97%

Supplier: Ambeed

Warning

A141691-1G A141691-5G
103834-076EA 96 USD
103834-076 103803-040
5-Fluoro-3-iodo-1H-indazole 97%
5-Fluoro-3-iodo-1H-indazole

Caution: Keep in a dark place, inert atmosphere, 2…8 °C

Formula: C₇H₄FIN₂
Boiling Pt: 361.4 °C (760 mm Hg)
Flash Pt: 172.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD06739145
CAS Number: 858629-06-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR