5-Bromo-2-methoxypyrimidine 98%

Supplier: Ambeed

Warning

A116042-1G A116042-5G
103824-526EA 13 USD
103824-526 103798-662
5-Bromo-2-methoxypyrimidine 98%
5-Bromo-2-methoxypyrimidine

Caution: Sealed in dry, at room temperature.

Formula: C₅H₅BrN₂O
Melting Pt: 55.5…59.5 °C
Flash Pt: 110.9 °C
Storage Temperature: Ambient
MDL Number: MFCD04973351
CAS Number: 14001-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR