5-Bromo-2-chloro-N-isopropylbenzamide 98%

Supplier: Ambeed
Warning

A940581-5G A940581-1G
103841-428EA 86.54 USD
103841-428 76845-698
5-Bromo-2-chloro-N-isopropylbenzamide 98%
5-Bromo-2-chloro-N-isopropylbenzamide

Caution: for Research Use Only

Formula: C₁₀H₁₁BrClNO
MW: 276.5600 g/mol
Melting Pt: 90…92 °C at N/A mmHg
Storage Temperature: Ambient
MDL Number: MFCD05853708
CAS Number: 849898-48-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR