5-Bromo-1,4-dimethyl-1H-1,2,3-triazole ≥95%

Supplier: Ambeed

Warning

A510641-1G
103828-388EA 1461 USD
103828-388
5-Bromo-1,4-dimethyl-1H-1,2,3-triazole ≥95%
5-Bromo-1,4-dimethyl-1H-1,2,3-triazole

Caution: Inert atmosphere, 2…8 °C.

Formula: C₄H₆BrN₃
MW: 176.0200 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD14706833
CAS Number: 1040275-55-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR