5-Amino-2,4,6-triiodoisophthalic acid 98%

Supplier: Ambeed
Warning

A115239-5G A115239-10G A115239-100G A115239-25G A115239-500G A115239-5G
76766-122EA 162.74 USD
76766-122 103824-380 77311-194 76766-120 76766-116 76766-118
5-Amino-2,4,6-triiodoisophthalic acid 98%
5-Amino-2,4,6-triiodoisophthalic acid

Caution: for Research Use Only

Formula: C₈H₄I₃NO₄
MW: 558.84 g/mol
Boiling Pt: 539.4 °C (760 mm Hg)
Melting Pt: 265…270 °C
Flash Pt: 280 °C
Storage Temperature: Ambient
MDL Number: MFCD00190167
CAS Number: 35453-19-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR