5,6-Dimethoxy-1-indanone 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
5,6-Dimethoxy-2,3-dihydro-1H-inden-1-one
A120912-5G
A120912-500G
A120912-25G
A120912-100G
A120912-10G
A120912-5G
76797-342EA
13.39
USD
76797-342
103830-822
76797-340
76797-344
76797-338
5,6-Dimethoxy-1-indanone 98%
5,6-Dimethoxy-1-indanone
Caution: for Research Use Only
Formula:
C₁₁H₁₂O₃ MW: 192.21 g/mol Boiling Pt: 339.7 °C (760 mm Hg) Melting Pt: 118…120 °C Flash Pt: 150.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003790 CAS Number: 2107-69-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...