5,6-Dibromo-2,1,3-benzothiadiazole 97%

Supplier: Ambeed
Warning

Synonyms: 5,6-Dibromobenzo[c][1,2,5]thiadiazole

A175904-5G A175904-1G A175904-250MG
103845-410EA 154.62 USD
103845-410 77236-408 103807-098
5,6-Dibromo-2,1,3-benzothiadiazole 97%
5,6-Dibromo-2,1,3-benzothiadiazole

Caution: for Research Use Only

Formula: C₆H₂Br₂N₂S
MW: 293.9700 g/mol
Boiling Pt: 334.5±22.0 °C (760 mm Hg)
Melting Pt: 132…133 °C
Flash Pt: 156.1 °C
Storage Temperature: Ambient
MDL Number: MFCD20257910
CAS Number: 18392-81-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR