4-(1-Pyrrolidinyl)pyridine 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
4-Pyrrolidinopyridine
A302882-25G
A302882-10G
A302882-5G
A302882-100G
A302882-5G
A302882-500G
76838-200EA
15.45
USD
76838-200
77002-558
76838-202
77002-556
77002-554
103835-512
4-(1-Pyrrolidinyl)pyridine 98%
4-(1-Pyrrolidinyl)pyridine
Caution: for Research Use Only
Formula:
C₉H₁₂N₂ MW: 148.21 g/mol Boiling Pt: 263.1 °C (760 mm Hg) Melting Pt: 57…58 °C Flash Pt: 112 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006415 CAS Number: 2456-81-7 UN: 2923 ADR: 8,6.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...