4-Phenyl-3-butyn-2-one 97%

Supplier: Ambeed
Warning

A452100-10G A452100-1G A452100-5G A452100-5G A452100-1G
103827-794EA 27.69 USD
103827-794 76842-694 77004-318 76842-692 103838-864
4-Phenyl-3-butyn-2-one 97%
4-Phenyl-3-butyn-2-one

Caution: for Research Use Only

Formula: C₁₀H₈O
MW: 144.17 g/mol
Boiling Pt: 252.942 °C (760 mm Hg)
Melting Pt: 4 °C at N/A mmHg
Flash Pt: 95 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008776
CAS Number: 1817-57-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR