Methyl-1H-pyrrolo[3,2-c]pyridin-4-ylether ≥95%

Supplier: Ambeed

Warning

Synonyms: 4-Methoxy-1H-pyrrolo[3,2-c]pyridine

A374427-1G
103826-548EA 221 USD
103826-548
Methyl-1H-pyrrolo[3,2-c]pyridin-4-ylether ≥95%
Methyl-1H-pyrrolo[3,2-c]pyridin-4-ylether

Caution: Sealed in dry, at room temperature.

Formula: C₈H₈N₂O
MW: 148.17 g/mol
Boiling Pt: 304.1 °C (760 mm Hg)
Melting Pt: 90…91 °C
Flash Pt: 104.5 °C
Storage Temperature: Ambient
MDL Number: MFCD08690141
CAS Number: 944900-76-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR