4-Iodo-1H-pyrazole 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Iodopyrazole
A133653-500G
A133653-5G
A133653-1G
A133653-1G
A133653-10G
A133653-100G
77298-646EA
5
USD
77298-646
103801-894
76972-238
76972-236
103832-828
76972-234
4-Iodo-1H-pyrazole 97%
4-Iodo-1H-pyrazole
Caution: for Research Use Only
Formula:
C₃H₃IN₂ MW: 193.97 g/mol Boiling Pt: 291.8 °C (760 mm Hg) Melting Pt: 108…110 °C Flash Pt: 130.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005244 CAS Number: 3469-69-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...