4-Iodopicolinic acid 97%

Warning

Synonyms: 4-Iodopyridine-2-carboxylic acid

A294447-5G A294447-1G
103814-008EA 20.77 USD
103814-008 103835-148
4-Iodopicolinic acid 97%
4-Iodopicolinic acid

Caution: Keep in a dark place, inert atmosphere, store in freezer, under -20 °C

Formula: C₆H₄INO₂
MW: 249.01 g/mol
Boiling Pt: 376.2 °C (760 mm Hg)
Melting Pt: 164…166 °C
Storage Temperature: Freezer
MDL Number: MFCD03426666
CAS Number: 405939-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR