3-Hydroxyphthalic anhydride 97%

Supplier: Ambeed
Warning

Synonyms: 4-Hydroxyisobenzofuran-1,3-dione

A247195-5G A247195-1G A247195-25G A247195-5G A247195-10G A247195-250MG A247195-100G A247195-1G
77090-906EA 10.3 USD
77090-906 77090-908 77193-528 77193-526 103851-588 77193-530 103811-756 77090-910
3-Hydroxyphthalic anhydride 97%
3-Hydroxyphthalic anhydride

Caution: for Research Use Only

Formula: C₈H₄O₄
MW: 164.1150 g/mol
Flash Pt: 160.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00011557
CAS Number: 37418-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR