4-Hydroxy-3-(trifluoromethyl)benzaldehyde 98%

Supplier: Ambeed

Warning

A274156-5G A274156-1G
103853-112EA 173 USD
103853-112 103813-188
4-Hydroxy-3-(trifluoromethyl)benzaldehyde 98%
4-Hydroxy-3-(trifluoromethyl)benzaldehyde

Caution: Inert atmosphere, 2…8 °C.

Formula: C₈H₅F₃O₂
MW: 190.12 g/mol
Boiling Pt: 223.9 °C (760 mm Hg)
Melting Pt: 159…163 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01091006
CAS Number: 220227-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR