4-Hexylphenylboronic acid 97%

Supplier: Ambeed

Warning

Synonyms: 4-Hexylphenylboronic Acid

A153365-1G A153365-5G
103804-498EA 39 USD
103804-498 103842-670
4-Hexylphenylboronic acid 97%
4-Hexylphenylboronic acid

Caution: Inert atmosphere, 2…8 °C.

Formula: C₁₂H₁₉BO₂
MW: 206.09 g/mol
Boiling Pt: 342.6 °C (760 mm Hg)
Flash Pt: 161 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04039029
CAS Number: 105365-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR