4-Fluoro-1H-indazole 95%

Supplier: Ambeed
Warning

Synonyms: 4-Fluoroindazole

A315423-5G A315423-250MG A315423-1G
103825-066EA 16.15 USD
103825-066 103835-992 77167-202
4-Fluoro-1H-indazole 95%
4-Fluoro-1H-indazole

Caution: for Research Use Only

Formula: C₇H₅FN₂
MW: 136.1264 g/mol
Boiling Pt: 274.9 °C (760 mm Hg)
Melting Pt: 130.9…133.7 °C
Storage Temperature: Ambient
MDL Number: MFCD07371561
CAS Number: 341-23-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR