4-Decylaniline 97%

Supplier: Ambeed

Warning

A300838-5G A300838-1G
103814-284EA 29 USD
103814-284 103835-434
4-Decylaniline 97%
4-Decylaniline

Caution: Keep in a dark place, inert atmosphere, room temperature

Formula: C₁₆H₂₇N
MW: 233.4 g/mol
Boiling Pt: 364.4 °C (760 mm Hg)
Melting Pt: 25 °C
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00007918
CAS Number: 37529-30-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR