4-Chloro-3-iodobenzoic acid 97%

Warning

A270746-5G
103852-918EA 12.69 USD
103852-918
4-Chloro-3-iodobenzoic acid 97%
4-Chloro-3-iodobenzoic acid

Caution: Keep in a dark place, sealed in dry, at 2…8°C

Formula: C₇H₄ClIO₂
Boiling Pt: 371.7 °C (760 mm Hg)
Melting Pt: 222…224 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00079732
CAS Number: 42860-04-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR