2-Amino-4-bromoquinoline 97%

Supplier: Ambeed

Warning

Synonyms: 4-Bromoquinolin-2-amine

A119882-1G
103799-854EA 453 USD
103799-854
2-Amino-4-bromoquinoline 97%
2-Amino-4-bromoquinoline

Caution: Keep in a dark place, inert atmosphere, 2…8 °C

Formula: C₉H₇BrN₂
Boiling Pt: 359 °C (760 mm Hg)
Melting Pt: 137…138 °C
Flash Pt: 171 °C
Storage Temperature: Refrigerator
CAS Number: 36825-32-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR