4-(Bromomethyl)benzenesulfonamide 96%

Supplier: Ambeed

Warning

Synonyms: 4-Bromomethylbenzenesulfonamide

A344468-1G A344468-5G
103836-886EA 395 USD
103836-886 103825-906
4-(Bromomethyl)benzenesulfonamide 96%
4-(Bromomethyl)benzenesulphonamide

Caution: Inert atmosphere, 2…8 °C.

Formula: C₇H₈BrNO₂S
MW: 250.1100 g/mol
Boiling Pt: 386.4 °C (760 mm Hg)
Melting Pt: 191…192 °C
Flash Pt: 187.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08752696
CAS Number: 40724-47-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR