2-Bromo-5-(trifluoromethyl)anisole 95%
|
Warning
|
Synonyms:
4-Bromo-3-methoxybenzotrifluoride
A321332-1G
A321332-5G
103836-204EA
20.77
USD
103836-204
103825-270
2-Bromo-5-(trifluoromethyl)anisole 95%
2-Bromo-5-(trifluoromethyl)anisole
Caution: Sealed in dry, at room temperature.
Formula:
C₈H₆BrF₃O MW: 255.03 g/mol Boiling Pt: 225 °C (760 mm Hg) Flash Pt: 109 °C Storage Temperature: Ambient |
MDL Number:
MFCD04115974 CAS Number: 402-07-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...