4-Bromo-3-iodopyridine 97%
Supplier: Ambeed
Warning
A279330-5G A279330-1G
103813-466EA 82 USD
103813-466 103853-410
4-Bromo-3-iodopyridine 97%
4-Bromo-3-iodopyridine
Caution: Keep in a dark place, inert atmosphere, store in freezer, under -20 °C