4-Bromo-2-hydroxybenzonitrile 98%

Supplier: Ambeed

Warning

A265561-5G A265561-1G
103852-630EA 22 USD
103852-630 103812-736
4-Bromo-2-hydroxybenzonitrile 98%
4-Bromo-2-hydroxybenzonitrile

Caution: Inert atmosphere, room temperature

Formula: C₇H₄BrNO
MW: 198.0168 g/mol
Boiling Pt: 295.1 °C (760 mm Hg)
Melting Pt: 160 °C
Flash Pt: 132.3 °C
Storage Temperature: Ambient
MDL Number: MFCD07780675
CAS Number: 288067-35-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR