4-Amino-N-methylbenzamide 98%

Supplier: AMBEED, INC

Warning

A165207-1000G A165207-10G
76690-012EA 9 USD
76690-012 103808-954
4-Amino-N-methylbenzamide 98%
4-Amino-N-methylbenzamide

Caution: for Research Use Only

Formula: C₈H₁₀N₂O
MW: 150.1778 g/mol
Boiling Pt: 364.1 °C (760 mm Hg)
Melting Pt: 181 °C
Storage Temperature: Ambient
MDL Number: MFCD00665792
CAS Number: 6274-22-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR