4-Amino-2,5-xylenol 97%

Supplier: Ambeed

Warning

Synonyms: 4-Amino-2,5-dimethylphenol

A100740-1G A100740-5G
103795-602EA 22 USD
103795-602 103821-300
4-Amino-2,5-xylenol 97%
4-Amino-2,5-xylenol

Caution: Keep in a dark place, inert atmosphere, room temperature

Formula: C₈H₁₁NO
MW: 137.1800 g/mol
Boiling Pt: 288.4 °C (760 mm Hg)
Melting Pt: 242…243 °C
Storage Temperature: Ambient
MDL Number: MFCD00007873
CAS Number: 3096-71-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR