4,4'-Dicyanobenzhydrol 97%

Supplier: Ambeed
Warning

A200141-1G A200141-250MG A200141-5G A200141-1G
103817-166EA 8.08 USD
103817-166 103847-794 77085-850 77237-108
4,4'-Dicyanobenzhydrol 97%
4,4'-Dicyanobenzhydrol

Caution: for Research Use Only

Formula: C₁₅H₁₀N₂O
MW: 234.2500 g/mol
Boiling Pt: 473.5 °C (760 mm Hg)
Flash Pt: 240.1 °C
Storage Temperature: Ambient
MDL Number: MFCD07367966
CAS Number: 134521-16-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR