4,4'-Diacetylbiphenyl 98%

Supplier: Ambeed

Warning

A263636-5G
103852-520EA 7 USD
103852-520
4,4'-Diacetylbiphenyl 98%
4,4'-Diacetylbiphenyl

Caution: Sealed in dry, at room temperature.

Formula: C₁₆H₁₄O₂
MW: 238.29 g/mol
Melting Pt: 193…195 °C
Storage Temperature: Ambient
MDL Number: MFCD00017248
CAS Number: 787-69-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR