2-Nitrophenylboronic acid pinacol ester 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Nitrophenylboronic acid Pinacol ester
, 4,4,5,5-Tetramethyl-2-(2-nitrophenyl)-1,3,2-dioxaborolane
, 2-Nitrobenzeneboronic acid pinacol ester
, 1-Nitro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene
, 2-Nitrophenylboronic acid, pinacol ester
A404182-100G
A404182-5G
A404182-1G
A404182-10G
A404182-250MG
A404182-5G
A404182-25G
A404182-1G
103838-144EA
26.54
USD
103838-144
76841-624
76725-116
76841-622
103827-104
76725-112
77307-086
76725-114
2-Nitrophenylboronic acid pinacol ester 98%
2-Nitrophenylboronic acid pinacol ester
Caution: for Research Use Only
Formula:
C₁₂H₁₆BNO₄ MW: 249.07 g/mol Boiling Pt: 358.2 °C (760 mm Hg) Melting Pt: 42…44 °C Flash Pt: 170.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD02179447 CAS Number: 190788-59-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...