2-Amino-4-(2-pyridyl)thiazole 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-(Pyridin-2-yl)thiazol-2-amine
A230347-1G
A230347-5G
103819-540EA
61
USD
103819-540
103850-438
2-Amino-4-(2-pyridyl)thiazole 97%
2-Amino-4-(2-pyridyl)thiazole
Caution: Keep in a dark place, sealed in dry, at 2…8°C
Formula:
C₈H₇N₃S MW: 177.22 g/mol Boiling Pt: 383.6 °C (760 mm Hg) Flash Pt: 185.8 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00460417 CAS Number: 30235-26-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...