4-(N,N-Diphenylamino)phenylboronic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-(Diphenylamino)benzeneboronic acid
, 4-(Diphenylamino)phenylboronic acid
A169779-100G
A169779-500G
A169779-5G
A169779-1G
A169779-5G
A169779-10G
77286-778EA
210
USD
77286-778
103806-406
77081-772
77286-780
103844-670
77286-782
4-(N,N-Diphenylamino)phenylboronic acid 98%
4-(N,N-Diphenylamino)phenylboronic acid
Caution: for Research Use Only
Formula:
C₁₈H₁₆BNO₂ MW: 289.14 g/mol Boiling Pt: 484.5 °C (760 mm Hg) Melting Pt: 110…115 °C Flash Pt: 246.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD06798117 CAS Number: 201802-67-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...