4-(4-Bromophenyl)morpholine 98%
Supplier: Ambeed
|
Warning
|
A257179-1G
A257179-1G
A257179-5G
A257179-100G
A257179-10G
A257179-5G
A257179-25G
77091-664EA
5.15
USD
77091-664
77098-754
77304-694
77098-752
77091-666
103852-158
103812-290
4-(4-Bromophenyl)morpholine 98%
4-(4-Bromophenyl)morpholine
Caution: for Research Use Only
Formula:
C₁₀H₁₂BrNO Boiling Pt: 341.2 °C (760 mm Hg) Melting Pt: 114…118 °C Storage Temperature: Ambient |
MDL Number:
MFCD04112483 CAS Number: 30483-75-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...