2-Hydroxy-4-pyridineboronic acid pinacol ester 97%
Supplier: Ambeed
Warning
Synonyms: 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2(1H)-one
A424981-5G A424981-1G
103827-452EA 256 USD
103827-452 103838-500
2-Hydroxy-4-pyridineboronic acid pinacol ester 97%
2-Hydroxy-4-pyridineboronic acid pinacol ester
Caution: Inert atmosphere, store in freezer, under –20 °C.