4-(Morpholinomethyl)phenylboronic acid pinacol ester 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-[4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]morpholine
, 4-(4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)morpholine
, 4,4,5,5-Tetramethyl-2-[4-(morpholinomethyl)phenyl]-1,3,2-dioxaborolane
, 4-(4-Morpholinylmethyl)benzeneboronic acid pinacol ester
, 4-(Morpholinomethyl)phenylboronic acid Pinacol ester
A313878-250MG
A313878-1G
A313878-10G
A313878-5G
A313878-5G
A313878-1G
103835-934EA
20.77
USD
103835-934
77306-102
103825-020
77290-178
77290-180
76838-748
4-(Morpholinomethyl)phenylboronic acid pinacol ester 95%
4-(Morpholinomethyl)phenylboronic acid pinacol ester
Caution: for Research Use Only
Formula:
C₁₇H₂₆BNO₃ MW: 303.20 g/mol Boiling Pt: 400.8 °C (760 mm Hg) Melting Pt: 76…78 °C Flash Pt: 196.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD04974052 CAS Number: 364794-79-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...