3-tert-Butylphenylboronic acid 95%
Supplier: Ambeed
|
Warning
|
A205105-1G
A205105-1G
A205105-5G
A205105-5G
77302-938EA
56
USD
77302-938
103817-598
103848-262
77086-462
3-tert-Butylphenylboronic acid 95%
3-tert-Butylphenylboronic acid
Caution: for Research Use Only
Formula:
C₁₀H₁₅BO₂ MW: 178.0400 g/mol Boiling Pt: 304.4 °C (760 mm Hg) Melting Pt: 111…113 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD11110647 CAS Number: 560132-24-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...