3'-Methoxypropiophenone 98%

Supplier: Ambeed
Warning

A248849-10G A248849-5G A248849-500G A248849-25G A248849-1G A248849-1G A248849-100G
77091-020EA 5.15 USD
77091-020 103851-678 103811-844
3'-Methoxypropiophenone 98%
3'-Methoxypropiophenone

Caution: for Research Use Only

Formula: C₁₀H₁₂O₂
MW: 164.2011 g/mol
Boiling Pt: 259 °C (760 mm Hg)
Flash Pt: 98.9 °C
Storage Temperature: Ambient
MDL Number: MFCD08458823
CAS Number: 37951-49-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR