3-Iodophthalic acid 95%

Supplier: Ambeed
Warning

A268614-1G A268614-5G A268614-100G A268614-5G A268614-1G A268614-25G A268614-10G
77092-580EA 21.63 USD
77092-580 77238-684 77238-686 77305-008 77092-578 103852-824 103812-922
3-Iodophthalic acid 95%
3-Iodophthalic acid

Caution: for Research Use Only

Formula: C₈H₅IO₄
MW: 292.0300 g/mol
Boiling Pt: 426.3 °C (760 mm Hg)
Melting Pt: 178…182 °C
Storage Temperature: Ambient
MDL Number: MFCD00275568
CAS Number: 6937-34-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR